1,3,5-Tribromobenzene ≥98%

140066-25G 140066-100G
220035-897EA 72.15 USD
220035-897 220035-896
1,3,5-Tribromobenzene ≥98%
1,3,5-Tribromobenzene
Formula: C₆H₃Br₃
MW: 314.8 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 122 - 123 °C
Density: 2.637 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR