Benzoyl bromide ≥97%

139726-250G 139726-50G
220036-697EA 998.31 USD
220036-697 220036-698
Benzoyl bromide ≥97%
Benzoyl bromide
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.527 to 1.5564 g/cm³ (10 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000112
CAS Number: 618-32-6
EINECS: 210-544-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR