N,N,N-Trimethylanilinium tribromide

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

139718-25G 139718-100G
220036-696EA 53.17 USD
220036-696 220036-695
N,N,N-Trimethylanilinium tribromide
N,N,N-Trimethylanilinium tribromide
Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
Melting Pt: 115…116 °C
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR