2,4-Dichlorobenzoic acid ≥98%

Warning

139572-5G 139572-100G
220036-691EA 37.69 USD
220036-691 220036-690
2,4-Dichlorobenzoic acid ≥98%
2,4-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 158 - 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002414
CAS Number: 50-84-0
EINECS: 200-067-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR