4-Fluorobenzonitrile ≥99%

Warning

Synonyms: p-Fluorobenzonitrile

139416-25G 139416-5G
220036-672EA 60.31 USD
220036-672 220036-673
4-Fluorobenzonitrile ≥99%
4-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 37 - 40 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001812
CAS Number: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR