3,5-Dichlorobenzaldehyde ≥97%

Warning

139408-1G 139408-5G
220035-971EA 38.77 USD
220035-971 220036-671
3,5-Dichlorobenzaldehyde ≥97%
3,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 215 - 222 °C (7 -rr)
Melting Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003352
CAS Number: 10203-08-4
EINECS: 233-499-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR