(4-Chlorophenyl)acetic acid ≥99%

Warning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

139262-100G 139262-10G
220035-959EA 96.49 USD
220035-959 220035-960
(4-Chlorophenyl)acetic acid ≥99%
(4-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 101 - 104 °C
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR