Isobutyryl chloride ≥98%

139122-100G 139122-5G 139122-500G
220036-663EA 43.62 USD
220036-663 220036-665 220036-664
Isobutyryl chloride ≥98%
Isobutyryl chloride
Formula: C₄H₇ClO
MW: 106.55 g/mol
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR