1,3,5,7-Cyclooctatetraene

Synonyms: Cyclooctatetraene, COT

138924-5G 138924-1G
220036-590EA 247.15 USD
220036-590 220036-589
1,3,5,7-Cyclooctatetraene
1,3,5,7-Cyclooctatetraene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: -7 °C
Density: 0.925 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004161
CAS Number: 629-20-9
EINECS: 211-080-3
UN: 2358
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR