1,3,5-Trimethoxybenzene ≥99%

Synonyms: O,O,O-1,3,5-Trimethylresorcinol

138827-10G 138827-250G 138827-50G
220036-585EA 37.26 USD
220036-585 220036-586 220036-587
1,3,5-Trimethoxybenzene ≥99%
1,3,5-Trimethoxybenzene
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008385
CAS Number: 621-23-8
EINECS: 210-673-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR