(4-Bromophenyl)acetic acid ≥98%

Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

138673-25G 138673-100G
220036-578EA 73.88 USD
220036-578 220036-577
(4-Bromophenyl)acetic acid ≥98%
(4-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 94…98 °C
MDL Number: MFCD00004342
CAS Number: 1878-68-8
EINECS: 217-523-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR