1-Chloro-2,4-dinitrobenzene ≥97%

Danger

138630-100G 138630-5G 138630-500G
220036-574EA 58.58 USD
220036-574 220036-576 220036-575
1-Chloro-2,4-dinitrobenzene ≥97%
1-Chloro-2,4-dinitrobenzene
Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007075
CAS Number: 97-00-7
EINECS: 202-551-4
UN: 3441
ADR: 6.1,II
Merck Index: 13,02155

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR