2,5-Dimethylbenzoic acid ≥95%

Warning

138177-10G 138177-50G
220035-555EA 523.94 USD
220035-555 220035-556
2,5-Dimethylbenzoic acid ≥95%
2,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 131 - 135 °C
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002482
CAS Number: 610-72-0
EINECS: 210-235-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR