2,3-Dimethylbenzoic acid ≥98%

Warning

138150-25G 138150-5G
220035-552EA 136.77 USD
220035-552 220035-553
2,3-Dimethylbenzoic acid ≥98%
2,3-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 143…147 °C
Density: 1.283 g/cm³ (20 °C)
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR