3,5-Xylidine ≥98%

Synonyms: 3,5-Dimethylaniline, 5-Amino-m-xylene

137863-500ML 137863-100ML
220036-476EA 270.31 USD
220036-476 220036-475
3,5-Xylidine ≥98%
3,5-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 9.8 °C
Density: 0.9706 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR