2,4-Di-tert-butylphenol ≥99%

Warning

Synonyms: 1,3-Di(dimethylethyl)-4-hydroxybenzene

137731-100G 137731-500G 137731-5G
220036-052EA 35.65 USD
220036-052 220036-053 220036-054
2,4-Di-tert-butylphenol ≥99%
2,4-Di-tert-butylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 53 - 56 °C
Density: 0.887 g/cm³ (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00008828
CAS Number: 96-76-4
EINECS: 202-532-0
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR