2,4,6-Tribromophenol ≥99%

Warning

137715-100G 137715-5G
220036-049EA 52.23 USD
220036-049 220036-050
2,4,6-Tribromophenol ≥99%
2,4,6-Tribromophenol
Formula: C₆H₃Br₃O
MW: 330.8 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 91…92 °C
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002150
CAS Number: 118-79-6
EINECS: 204-278-6
Merck Index: 12,09744

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR