2-Bromoethyl acetate ≥97%

Synonyms: Acetic acid (2-bromoethyl) ester

137685-500G 137685-100G 137685-5G
220036-047EA 38.54 USD
220036-047 220036-046 220036-048
2-Bromoethyl acetate ≥97%
2-Bromoethyl acetate
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1.4623 g/cm³ (20 °C)
MDL Number: MFCD00000235
CAS Number: 927-68-4
EINECS: 213-159-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR