2-Bromobenzoic acid ≥97%

Warning

Synonyms: o-Bromobenzoic Acid

137677-100G 137677-25G
220036-044EA 69.57 USD
220036-044 220036-045
2-Bromobenzoic acid ≥97%
2-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 144…147 °C
Density: 1.929 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR