3,4-Dimethylanisole ≥99%

137499-250G 137499-50G
220036-459EA 451.23 USD
220036-459 220036-460
3,4-Dimethylanisole ≥99%
3,4-Dimethylanisole
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 200 °C (1013 hPa)
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00008396
CAS Number: 4685-47-6
EINECS: 225-142-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR