5-Methyl-2-furaldehyde

Synonyms: 5-Methylfurfural, 5-Methyl-2-Furaldehyde

137316-100G 137316-25G 137316-5G
220036-443EA 135.69 USD
220036-443 220036-444 220036-445
5-Methyl-2-furaldehyde
5-Methyl-2-furaldehyde
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 89 °C (1013 hPa)
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR