1-Phenyl-2-butanone ≥98%

Synonyms: 1-Phenylbutan-2-one, Benzyl ethyl ketone

137227-25G 137227-5G
220036-366EA 577.23 USD
220036-366 220036-367
1-Phenyl-2-butanone ≥98%
1-Phenyl-2-butanone
Formula: C₆H₅C(O)CH₂CH₂CH₃
MW: 148.2 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 5 °C
Density: 0.989 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009315
CAS Number: 1007-32-5
EINECS: 213-752-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR