4,4-Dimethyl-2-pentanone ≥99%

Synonyms: 4,4-Dimethylpentan-2-one

136875-5G 136875-25G
220036-357EA 188.46 USD
220036-357 220036-356
4,4-Dimethyl-2-pentanone ≥99%
4,4-Dimethyl-2-pentanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: -64 °C
Density: 0.808 g/cm³ (20 °C)
MDL Number: MFCD00008854
CAS Number: 590-50-1
EINECS: 209-685-2
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR