(±)-3,3-Dimethyl-2-butanol ≥98%

Synonyms: Pinacolyl alcohol

136824-25G 136824-100G
220036-352EA 160.46 USD
220036-352 220036-351
(±)-3,3-Dimethyl-2-butanol ≥98%
(±)-3,3-Dimethyl-2-butanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: 5 °C
Density: 0.8347 g/cm³ (0 °C)
MDL Number: MFCD00004522
CAS Number: 464-07-3
EINECS: 207-347-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR