2-Methyl-3-buten-2-ol ≥98%

Synonyms: 3-Hydroxy-3-methyl-1-butene

136816-100ML 136816-500ML 136816-1L
220035-698EA 33.28 USD
220035-698 220035-700 220035-699
2-Methyl-3-buten-2-ol ≥98%
2-Methyl-3-buten-2-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.824 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR