Dimethyl malonate ≥98%

136441-1KG 136441-250G
220035-677EA 119.54 USD
220035-677 220035-678
Dimethyl malonate ≥98%
Dimethyl malonate
Formula: C₅H₈O₄
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: -62 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR