2-Methyl-4(5)-nitroimidazole

Synonyms: 2-Methyl-4-nitro-1H-imidazole, 2-Methyl-5-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole, 2-Methyl-4-nitroimidazole

136255-50G 136255-250G
220036-341EA 52.12 USD
220036-341 220036-340
2-Methyl-4(5)-nitroimidazole
2-Methyl-4(5)-nitroimidazole
Formula: C₄H₅N₃O
MW: 127.10 g/mol
MDL Number: MFCD00005191
CAS Number: 696-23-1
EINECS: 211-790-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR