trans-Chalcone ≥97%

Synonyms: trans-Benzylidenacetophenone, (E)-Chalcone, trans-Benzalacetophenone, (E)-1,3-Diphenylprop-2-en-1-one

136123-100G 136123-5G
220035-947EA 64.62 USD
220035-947 220035-948
trans-Chalcone ≥97%
trans-Chalcone
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 55 - 57 °C
MDL Number: MFCD00003082
CAS Number: 614-47-1
EINECS: 210-383-8
Merck Index: 13,02028
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f708bafe7d3c2e194d3e9bf5f410a2c30afb756&vid=aa90a6b415f359c4551772765e81cbdff2cce599&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR