2-Chloro-5-nitrobenzonitrile ≥99%

135712-100G 135712-25G
220036-330EA 292.92 USD
220036-330 220036-331
2-Chloro-5-nitrobenzonitrile ≥99%
2-Chloro-5-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Boiling Pt: 119…122 °C (0.6 mmHg)
Melting Pt: 108 °C
MDL Number: MFCD00007292
CAS Number: 16588-02-6
EINECS: 240-643-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR