2-Bromo-2-methylpropane ≥98%

Danger

Synonyms: tert-Butyl bromide

135615-5G 135615-500G 135615-100G
220036-321EA 38.02 USD
220036-321 220036-320 220036-319
2-Bromo-2-methylpropane ≥98%
2-Bromo-2-methylpropane
Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 73 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.2125 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00000125
CAS Number: 507-19-7
EINECS: 208-065-9
UN: 2342
ADR: 3,II
Merck Index: 13,01554

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR