4-Chlorobenzoic acid ≥99%

Warning

Synonyms: p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid

135585-50G 135585-250G
220036-318EA 43.62 USD
220036-318 220036-317
4-Chlorobenzoic acid ≥99%
4-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 123 °C
Density: 1.535 g/cm³ (20 °C)
Flash Pt: 238 °C
Storage Temperature: Ambient
MDL Number: MFCD00002531
CAS Number: 74-11-3
EINECS: 200-805-9
Merck Index: 13,02144

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR