2-Chlorobenzoic acid ≥98%

Warning

135577-100G 135577-5G 135577-500G
220036-314EA 39.85 USD
220036-314 220036-316 220036-315
2-Chlorobenzoic acid ≥98%
2-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1.544 g/cm³ (20 °C)
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00002412
CAS Number: 118-91-2
EINECS: 204-285-4
Merck Index: 13,02143

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR