3-Phenylpropionic acid ≥99%

Warning

Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid

135232-5G 135232-100G
220036-303EA 21 USD
220036-303 220036-302
3-Phenylpropionic acid ≥99%
3-Phenylpropionic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 46…47 °C
Density: 1.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR