Boron trifluoride - methanol complex 50% in methanol

Danger

134821-50ML 134821-250ML 134821-1L
220036-276EA 74.77 USD
220036-276 220036-275 220036-274
Boron trifluoride - methanol complex 50% in methanol
Boron trifluoride - methanol complex
Formula: CH₄BF₃O
MW: 99.85 g/mol
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR