2-Acetylnaphthalene

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

134775-5G 134775-100G
220036-266EA 142.15 USD
220036-266 220036-265
2-Acetylnaphthalene
2-Acetylnaphthalene
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.1171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR