Ethyl bromoacetate ≥98%, Reagent Grade

Danger

Synonyms: Bromoacetic ethyl ester

133973-100G 133973-500G 133973-5G
220035-880EA 44.05 USD
220035-880 220035-881 220035-882
Ethyl bromoacetate ≥98%, Reagent Grade
Ethyl bromoacetate
Formula: BrCH₂COOCH₂CH₃
MW: 167 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: -38 °C
Density: 1.5 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000191
CAS Number: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR