2-Amino-4-chlorobenzothiazole

133434-25G 133434-5G
220036-192EA 57.08 USD
220036-192 220036-193
2-Amino-4-chlorobenzothiazole
2-Amino-4-chlorobenzothiazole
Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
MDL Number: MFCD00005791
CAS Number: 19952-47-7
EINECS: 243-439-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR