1,2-Dibromotetrachloroethane ≥97%

Warning

133396-100G 133396-25G
220036-189EA 137.85 USD
220036-189 220036-190
1,2-Dibromotetrachloroethane ≥97%
1,2-Dibromotetrachloroethane
Formula: BrCl₂CCBrCl₂
MW: 325.64 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 83 °C
Density: 2.713 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000780
CAS Number: 630-25-1
EINECS: 211-136-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR