(±)-2-Methyl-1-butanol ≥99%

Danger

133051-1L 133051-100ML
220036-142EA 88.31 USD
220036-142 220036-141
(±)-2-Methyl-1-butanol ≥99%
(±)-2-Methyl-1-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004743
CAS Number: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3,III
Merck Index: 12,06109

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR