4,4'-Isopropylidenediphenol ≥97%

Synonyms: Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propane

133027-500G 133027-2KG
220036-136EA 47.2 USD
220036-136 220035-673
4,4'-Isopropylidenediphenol ≥97%
4,4'-Isopropylidenediphenol
Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 155 °C
Density: 1.205 g/cm³ (20 °C)
Flash Pt: 227 °C
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
Merck Index: 13,01296

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR