4-Nitro-2,6-xylenol ≥98%

Synonyms: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol

132713-1G 132713-10G
220036-128EA 45 USD
220036-128 220036-127
4-Nitro-2,6-xylenol ≥98%
4-Nitro-2,6-xylenol
Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007339
CAS Number: 2423-71-4
EINECS: 219-353-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR