3,5-Dimethoxyphenol ≥99%

Warning

132632-10G 132632-50G
220035-514EA 109.85 USD
220035-514 220035-515
3,5-Dimethoxyphenol ≥99%
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00008388
CAS Number: 500-99-2
EINECS: 207-917-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR