1,2-Diaminocyclohexane cis- and trans-mixture ≥99%

Synonyms: 1,2-Diaminocyclohexane, 1,2-Cyclohexanediamine

132551-1L 132551-50ML 132551-250ML 132551-18L
220036-119EA 252 USD
220036-119 220036-121 220036-120 220036-118
1,2-Diaminocyclohexane cis- and trans-mixture ≥99%
1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0.93 g/cm³ (20 °C)
MDL Number: MFCD00001491
CAS Number: 694-83-7
EINECS: 211-776-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR