4-Bromobutyl phenyl ether ≥97%

Synonyms: 4-Phenoxybutyl bromide, 1-Bromo-4-phenoxybutane, (4-Bromobutoxy)benzene

132543-25G 132543-100G
220036-117EA 96.15 USD
220036-117 220036-116
4-Bromobutyl phenyl ether ≥97%
4-Bromobutyl phenyl ether
Formula: (C₆H₅)O(CH₂)₄Br
MW: 229.12 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 141 °C
MDL Number: MFCD00000262
CAS Number: 1200-03-9
EINECS: 214-849-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR