Zinc(II) acetylacetonate hydrate

Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II) hydrate, Zinc acetylacetonate xhydrate, Zinc (II) 2,4-pentanedionate hydrate, Zinc (II) bis(2,4-pentanedionato-O,O') hydrate, Zinc acetylacetonate hydrate

132306-100G 132306-500G
220036-096EA 148.77 USD
220036-096 220036-097
Zinc(II) acetylacetonate hydrate
Zinc(II) acetylacetonate hydrate
Formula: C₁₀H₁₄O₄Zn·H₂O
MW: 281.62 g/mol
MDL Number: MFCD00149060
CAS Number: 108503-47-5
EINECS: 237-860-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR