2-Vinylpyridine

132292-5ML 132292-500ML 132292-100ML
220036-095EA 45.51 USD
220036-095 220036-094 220036-093
2-Vinylpyridine
2-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.9735 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Freezer
MDL Number: MFCD00006355
CAS Number: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR