10-Undecenal ≥95%

Synonyms: Undecylenic aldehyde, Undec-10-enal, 10-Undecenyl aldehyde, 10-Undecenal

132276-100G 132276-25G
220036-091EA 150 USD
220036-091 220036-092
10-Undecenal ≥95%
10-Undecenal
Formula: C₁₁H₂₀O
MW: 168.28 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.8467 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007032
CAS Number: 112-45-8
EINECS: 203-973-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR