2,4,6-Trimethylaniline ≥98%

Warning

132179-25ML 132179-100ML
220036-083EA 48.89 USD
220036-083 220036-082
2,4,6-Trimethylaniline ≥98%
2,4,6-Trimethylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: -5 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007740
CAS Number: 88-05-1
EINECS: 201-794-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR