2,4,5-Trimethoxybenzaldehyde ≥98%

132152-100G 132152-25G
220036-079EA 186.92 USD
220036-079 220036-080
2,4,5-Trimethoxybenzaldehyde ≥98%
2,4,5-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003312
CAS Number: 4460-86-0
EINECS: 224-713-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR