Sodium trifluoroacetate ≥98%

Synonyms: Sodium 2,2,2-trifluoroacetate

132101-100G 132101-25G
220036-077EA 78.92 USD
220036-077 220036-078
Sodium trifluoroacetate ≥98%
Sodium trifluoroacetate
Formula: C₂F₃NaO₂
MW: 136.01 g/mol
Melting Pt: 207 - 209 °C
MDL Number: MFCD00013217
CAS Number: 2923-18-4
EINECS: 220-879-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR